Learn More
Invitrogen™ WBP4 Polyclonal Antibody

Description
WBP4 Polyclonal Antibody for ELISA, Western Blot
Specifications
Specifications
| Antigen | WBP4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | WBP4 |
| Gene Accession No. | Q5HZF2 |
| Gene Alias | AW545037; BB101031; domain-containing binding protein 4; FBP21; Fnbp21; formin binding protein 21; formin-binding protein 21; hypothetical protein LOC553723; MGC117310; Wbp4; WBP-4; WW domain binding protein 4; WW domain binding protein 4 (formin binding protein 21); WW domain-binding protein 4; WW domain-binding protein 4-like protein; WW domain-containing binding protein 4; WW domain-containing-binding protein 4; zgc:112384; zgc:112384 protein |
| Gene Symbols | WBP4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.