Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDFY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567604
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567604 Supplier Invitrogen™ Supplier No. PA567604
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDFY2 Polyclonal Antibody for Western Blot, IHC (P)

The WDFY2 gene encodes a protein that contains two WD domains and an FYVE zinc finger region. The function of this gene is unknown. An alternatively spliced transcript variant of this gene may exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDFY2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene WDFY2
Gene Accession No. Q8BUB4, Q96P53
Gene Alias 5830485M08; B130024L21Rik; ProF; propeller-FYVE protein; WD repeat and FYVE domain containing 2; WD repeat and FYVE domain-containing protein 2; WD40 and FYVE domain containing 2; WD40- and FYVE domain-containing protein 2; WDF2; WDFY2; ZFYVE22; zinc finger FYVE domain-containing protein 22
Gene Symbols WDFY2
Host Species Rabbit
Immunogen Fusion protein of human WDFY2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115825, 268752
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.