Learn More
Invitrogen™ WDR1 Monoclonal Antibody (5C11C8)

Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Immunogen sequence is different from the related and rat sequences by one amino acid. Positive Control - WB: human U87 whole cell, human A549 whole cell, mouse lung tissue, mouse liver tissue. IHC: human liver cancer tissue, human colorectal adenocarcinoma tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | WDR1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 5C11C8 |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and no preservative |
| Gene | WDR1 |
| Gene Accession No. | O75083, O88342 |
| Gene Alias | Actin-interacting protein 1; Aip1; D5Wsu185e; epididymis secretory protein Li 52; HEL-S-52; NORI-1; rede; WD repeat domain 1; WD repeat protein 1; WD repeat-containing protein 1; WD40 repeat protein 1; Wdr1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.