Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555892 Supplier Invitrogen™ Supplier No. PA555892
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NDLNELKPLV HSPHAIVRMK FLLLQQKYLE YLEDGKVLEA LQVLRCELTP LKYNTERIHV LSGYLMCSHA EDLRAKAEWE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 83%.

May be involved in MAPK pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR26
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR26
Gene Accession No. Q9H7D7
Gene Alias 1600024A01Rik; AA693241; AI447817; AU044014; C77982; CDW2; CUL4- and DDB1-associated WDR protein 2; FLJ21016; GID complex subunit 7 homolog; Gid7; MIP2; myocardial ischemic preconditioning upregulated protein 2; Myocardial ischemic preconditioning up-regulated protein 2; PRO0852; RGD1565589; WD repeat domain 26; WD repeat-containing protein 26; Wdr26
Gene Symbols WDR26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR26. Recombinant protein control fragment (Product #RP-94421).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80232
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.