Learn More
Invitrogen™ WDR44 Polyclonal Antibody

Description
Immunogen sequence: MRMKYNTEGR VSPSPSQESL SSSKSDTDTG VCSGTDEDPD DKNAPFRQRP FCKYKGHTAD LLDLSWSKNY FLLSSSMDKT VR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Specifications
Specifications
| Antigen | WDR44 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | WDR44 |
| Gene Accession No. | Q5JSH3 |
| Gene Alias | RAB11BP; rabphilin-11; RPH11; WD repeat domain 44; WD repeat-containing protein 44; WDR44 |
| Gene Symbols | WDR44 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.