Learn More
Invitrogen™ WDR45B Polyclonal Antibody

Description
The antibody detects endogenous levels of total WDR45B protein.
Specifications
Specifications
| Antigen | WDR45B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | WDR45B |
| Gene Accession No. | Q5MNZ6 |
| Gene Alias | 0610008N23Rik; AA408735; D16Bwg0193e; RGD1305141; WD repeat domain 45B; WD repeat domain phosphoinositide-interacting protein 3; WD repeat protein 45-like; WD repeat-containing protein 45B; WD repeat-containing protein 45-like; Wdr45 like; WDR45B; Wdr45l; WDR45-like; WDR45-like protein; WIPI3; WIPI-3; WIPI49-like protein; zgc:56002 |
| Gene Symbols | WDR45B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.