Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR47 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555571
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555571 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555571 Supplier Invitrogen™ Supplier No. PA555571
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EHSVIKPPLG DSPGSLSRSK GEEDDKSKKQ FVCINILEDT QAVRAVAFHP AGGLYAVGSN SKTLRVCAYP DVIDPSAHET PKQPVV Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR47
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR47
Gene Accession No. O94967
Gene Alias KIAA0893; Nemitin; neuronal enriched MAP interacting protein; neuronal enriched MAP-interacting protein; WD repeat domain 47; WD repeat-containing protein 47; WDR47
Gene Symbols WDR47
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR47. Recombinant protein control fragment (Product #RP-94037).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22911
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.