Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR61 Monoclonal Antibody (OTI3C8), TrueMAB™, OriGene
SDP

Catalog No. 501681636
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1636 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1636 Supplier OriGene Supplier No. TA807591
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SKI8 is a component of the PAF and SKI complex which is composed of SKI2, SKI3 and SKI8. These are involved in transcriptional regulation and events downstream of RNA synthesis. Interaction of the SKI complex with SKI7 links the SKI complex to the exosome in order to perform mRNA degradation.

Specifications

Antigen WDR61
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI3C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene WDR61
Gene Accession No. Q9GZS3
Gene Alias REC14, SKI8
Gene Symbols WDR61
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 92-305 of human WDR61 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80349
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.