Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR83 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545085
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545085 Supplier Invitrogen™ Supplier No. PA545085
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EGALALALPV GSGVVQSLAY HPTEPCLLTA MGGSVQCWRE EAYEAEDGAG Sequence homology: Cow: 85%; Dog: 86%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 85%; Rabbit: 79%; Rat: 85%.

MORG1 is a member of the WD-40 protein family. The protein is proposed to function as a molecular scaffold for various multimeric protein complexes. The protein associates with several components of the extracellular signal-regulated kinase (ERK) pathway, and promotes ERK activity in response to serum or other signals. The protein also interacts with egl nine homolog 3 (EGLN3, also known as PHD3) and regulates expression of hypoxia-inducible factor 1, and has been purified as part of the spliceosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR83
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene WDR83
Gene Accession No. Q9BRX9
Gene Alias 1500041N16Rik; MAPK cascade scaffold protein; MAPK organizer 1; MGC4238; Mitogen-activated protein kinase organizer 1; MORG1; RGD1306947; WD repeat domain 83; WD repeat domain containing 83; WD repeat domain-containing protein 83; WD repeat protein Morg1; WDR83
Gene Symbols WDR83
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human WDR83 (aa 266-315).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84292
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.