Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wearwell™ Industrial WorkSafe™ Mat

Catalog No. 17986112 Shop All Wearwell Tennessee Mat Company Inc. Products
Change view
Click to view available options
Dimensions (L x W):
40 x 28 in.
64 x 40 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17-986-112 40 x 28 in.
17-986-114 64 x 40 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-986-112 Supplier Wearwell™ Supplier No. 477.78X28X40BOR

Extra-thick workstation mats withstand many industrial chemicals

  • Designed for metal working and other rough industrial areas where performance and productivity count
  • These extra thick and extra buoyant mats provide long-term comfort
  • The CFR (mineral oil cutting fluid resistant) compound withstands many non-petroleum based industrial fluids
  • The bright yellow or orange, molded-in safety borders define workstations while the mats textured surface adds traction in wet areas
  • In hazardously slippery areas, use the GritWorks version This high performance carbide grit coating offers 60% more traction than specified by OSHA in wet areas
  • 0.875 in. thick

Specifications

Resistance Grease-resistant
Material Rubber
Dimensions (L x W) 40 x 28 in.
Color Black/Orange
Thickness (English) 0.87 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.