Description
- No-wax vinyl surface for use in dry work areas
- Supercompression sponge base relieves overall body fatigue
- 0.08 in. (0.2cm) thick virgin vinyl permanently bonded to 0.38 in. (1cm) thick moisture-resistant, closed-cell sponge base
- Tough, nonporous surface resists abuse even in the most heavily used work areas
- Surface resists breakdown when exposed to most laboratory and industrial chemicals
- Edges safely beveled to prevent tripping and falling
Specifications
Specifications
| Color | Black |
| Dimensions (L x W) | 3 x 5 ft. |