Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaPhoenix™ Weather Wonders Deluxe Kit Components
SCI_ED

Catalog No. S39377KWW Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Product Type:
Cotton Swabs
Craft Stick
Fine Sand
Latch top Storage Container
Zipseal bag
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S39377KWW Fine Sand
S39387KPP Zipseal bag
S39381KWW Cotton Swabs
S39386KPP Craft Stick
S395513LG Latch top Storage Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S39377KWW
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement parts for Educational Kit

Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.

Specifications

Grade K to 5
For Use With (Application) Education Kit
Quantity 4 cup
Product Type Fine Sand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.