Learn More
Description
Weathered materials indicate the percent weight loss from the original material
Components:- Diesel Fuel #2 Standard
- Diesel Fuel #2 Standard: 25% Weathered
- Diesel Fuel #2 Standard: 50% Weathered
- Diesel Fuel #2 Standard: 75% Weathered
Specifications
Specifications
| CAS | 68334-30-5, 75-09-2 |
| CAS Max % | 99.5 |
| CAS Min % | 0.5 |
| Odor | Strong |
| Vapor Density | 2.93 (air = 1) |
| Color | Colorless |
| Melting Point | -96.7°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
