Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Weathered Gasoline Kit #2, Restek™
SDP

Catalog No. 06728572 Shop All Restek Corporation Products
Encompass_Preferred
Catalog No. 06-728-572 Supplier Restek Corporation Supplier No. 30100

missing translation for 'validMainframeMsg'

Preparation of these solutions are from a single-source (one-refinery) product. The weathered materials indicate the percent weight loss from the original material. Samples of regular and premium grade unleaded gasoline were collected, then blended in equal volumes.

Contains 1mL each of these mixtures.

30096: Unleaded Gasoline (ASTM Class 2 Accelerant)

  • Unleaded gasoline: unweathered (8006-61-9)

30097: Unleaded Gasoline (ASTM Class 2 Accelerant)

  • Unleaded gasoline: 25% weathered (8006-61-9)

30098: Unleaded Gasoline (ASTM Class 2 Accelerant)

  • Unleaded gasoline: 50% weathered (8006-61-9)

30099: Unleaded Gasoline (ASTM Class 2 Accelerant)

  • Unleaded gasoline: 75% weathered (8006-61-9)

Specifications

Description 0%, 25%, 50%, 75% Weathered
Quantity 1 mL
ShelfLife 6 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.