Learn More
Welch™ Standard Duty WOB-L™ Piston Vacuum Pump: Model 2546
Description
Includes
Includes convenient gauge read cut, pressure adjustment valve, protective separator jar, North america and Euro (Schuko) plug
For non-chemical applications
Wobbling piston wobbles providing air resistance on upward stroke expanding Teflon seal on piston and increases its efficiency for wobble action.
- Oil free dry pump powered by WOB-L piston
- Intake/Exhaust thread NPT: 1/4
Model 2546
- Free air displacement: CFM (L/min) at 60Hz: 1.6 (45)
- Ultimate pressure: 60 torr (80mbar)
- Maximum pressure: 100psig
- Maximum vacuum in Hg: 27.6
- Motor: 0.25 hp (190w)
- For pumping vapors of aqueous solutions including buffers but not for acidic, basic or organic vapors or gases
- 50Hz units supplied with CE marking
Vacuum filtration, Vacuum oven, Vacuum chamber/Roughing; Dessication; Aspiration; Automation; Cell culture; Glove box
Specifications
Specifications
| Horsepower | 0.25 hp |
| Shipping Weight (English) | 17 lb. |
| Shipping Weight (Metric) | 7.7 kg |
| Includes | Convenient gauge read cut, pressure adjustment valve, protective separator jar, North American 230V North American plug |
| Certifications/Compliance | CE |
| Electrical Requirements | 230 V 60 Hz |
| Model | 2546 |
| Weight (English) | 13.8 lb. |
| Weight (Metric) | 6.3 kg |
| Flow Rate | 2.3 CFM |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.