Learn More
Restek™ Weldment Removal Tool for EZ Twist Top Split/Splitless Injection Ports
SDP
Catalog No. 01001505 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
For Agilent GCs
Catalog No. For Use With (Equipment)
01-001-505 For Agilent GCs
1 options
Catalog No. 01-001-505 Supplier Restek™ Supplier No. 22728
For quick removal of hot weldment without fingers touching hot surfaces
  • Stays secure in tool for reattachment
  • For Agilent GCs
Specifications
Product Type Weldment Removal Tool
For Use With (Equipment) For Agilent GCs