Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Well Plate Accessories
GSA_VA

Catalog No. 60105400
Change view
Click to view available options
Product Type:
Automated Sample Concentrator
Block
Center Hole Cap
Deep Well Plate Vial
Mat Applicator
Microtiter Plate
Microtiter Plate Vial
Needle Kit with Fitting Tool
Rack
Sealing Tape Applicator
Square Well Plate Vial
Tube Tray
Volume (Metric) Working:
1000 μL
1100 μL
1500 μL
2200 μL
350 μL
500 μL
700 μL
Not Applicable
Well Shape:
Flat Bottom (F-well)
Not Applicable
Round
Round Bottom (U-well)
Square
Taper
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Volume (Metric) Working Well Shape
60-105-400 Microtiter Plate Vial 500 μL Taper
60-105-424 Deep Well Plate Vial 350 μL Taper
60-105-401 Microtiter Plate Vial 700 μL Taper
60-105-425 Square Well Plate Vial 1000 μL Round
60-105-403 Microtiter Plate Vial 1100 μL Round
60-105-405 Deep Well Plate Vial 1500 μL Round
03-252-446 Mat Applicator Not Applicable Not Applicable
03-252-445 Sealing Tape Applicator Not Applicable Not Applicable
60-105-432 Tube Tray Not Applicable Not Applicable
60-105-427 Center Hole Cap Not Applicable Not Applicable
60-105-429 Microtiter Plate 350 μL Flat Bottom (F-well)
60-105-402 Microtiter Plate Vial 700 μL Round
60-105-406 Square Well Plate Vial 1000 μL Square
60-105-416 Microtiter Plate 2200 μL Round Bottom (U-well)
60-105-434 Automated Sample Concentrator Not Applicable Not Applicable
60-105-435 Needle Kit with Fitting Tool Not Applicable Not Applicable
60-105-431 Block Not Applicable Round Bottom (U-well)
60-105-415 Rack Not Applicable Not Applicable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 60-105-400 Supplier Thermo Scientific™ Supplier No. 60180V100
Add to Cart
Add to Cart

Effectively seal all microplate formats to minimize evaporation and protect samples from contamination and spilling, ensure a consistent, reliable seal with all microplate formats with this easy-to-use, handheld applicator, and get significantly quicker dry-down times than standard methods, and remove most chromatography solvents in just minutes with easy to use 96-well sample concentrators.

Sealing Tape Applicator
Ensure a consistent, reliable seal with all microplate formats and all types of tape with this easy to use, handheld applicator for Thermo Scientific™ WebSeal Sealing Tapes. They are packaged two per box.

Sample Concentrators
Remove most chromatography solvents in just minutes. Simple to operate and maintain, Thermo Scientific™ WebSeal Sample Concentrators allow quick, easy solvent evaporation from 96 well microplates, offering significantly quicker dry-down times than standard methods such as vacuum ovens or centrifuges. These evaporators offer significant throughput advantages for laboratories looking to accelerate microplate sample preparation. Depending on your throughput requirements, choose either the manually operated MiniVap Concentrator or the programmable, robot-ready UltraVap.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Product Line Webseal
Color Clear
Product Type Microtiter Plate Vial
For Use With (Equipment) 96-well Microtiter Plates
Material Glass
No. of Wells 96
Volume (Metric) Working 500 μL
Total Volume (Metric) Well 500 μL
Well Shape Taper
Plate Compatibility 96-well Plates
Length (Metric) 6.5 mm
Width (Metric) 31.0 mm
Show More Show Less
What is the difference between the SureSTART Performance Level 1, Performance Level 2, and Performance Level 3 vials?

All glass vials found within the three different performance levels are made from Type 1 Class A Borosilicate glass. This type of glass has excellent chemical resistance properties that make it ideal for the containment or storage of acidic, neutral and alkaline samples/analytes. The vials also have high temperature resistance and are suitable for use at temperatures between -50 degrees C to 120 degrees C (although a rapid change in temperature of the products should be avoided).

The Performance Level 1 vials are designed to identify those which are suitable for simple routine, testing with limited budget. Performance Level 1 glass vials have a single wavy line.

For most analytical requirements, the Performance Level 2 products will be suitable for use with higher sensitivity methods while maintaining throughput. Performance Level 2 glass vials have two wavy lines.

For the most stringent analytical requirements, the Performance Level 3 products have advanced features which make them suitable for use with difficult samples in terms of chemistry and available sample volume. All Performance Level 3 products are made from 33-expansion glass and these vials have three wavy lines.

What happened to National, Chromacol, and other brands for chromatogenic workflow accessories?

Many of the products that were branded as National Scientific and Chromacol have been absorbed into the new Thermo Scientific SureSTART Collection, although there are still a few products that still exist within the marketplace.

For more information, please contact your local sales representative or complete the inquiries form at the bottom of the SureSTART Autosampler Vials and Caps page (https://www.thermofisher.com/us/en/home/industrial/chromatography/chromatography-consumables/autosampler-vials-caps-hplc-gc.html).

What is the SureSTART products portfolio for chromatographic workflows?

The SureSTART portfolio is a comprehensive list of vials, caps, inserts, well plates, mats and accessories that are suitable for use with chromatographic workflows. The SureSTART portfolio contains three different performance levels that have been quality checked to provide a robust and reliable product.

The SureSTART portfolio includes products that were previously found in our legacy brands such as National Scientific, Chromacol, and Sun-Sri.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.