Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Well Plate Accessories

Catalog No. 60105402
Change view
Click to view available options
Product Type:
Automated Sample Concentrator
Block
Center Hole Cap
Deep Well Plate Vial
Mat Applicator
Microtiter Plate
Microtiter Plate Vial
Needle Kit with Fitting Tool
Rack
Sealing Tape Applicator
Square Well Plate Vial
Tube Tray
Volume (Metric) Working:
1000 μL
1100 μL
1500 μL
2200 μL
350 μL
500 μL
700 μL
Not Applicable
Well Shape:
Flat Bottom (F-well)
Not Applicable
Round
Round Bottom (U-well)
Square
Taper
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Volume (Metric) Working Well Shape
60-105-402 Microtiter Plate Vial 700 μL Round
60-105-424 Deep Well Plate Vial 350 μL Taper
60-105-400 Microtiter Plate Vial 500 μL Taper
60-105-401 Microtiter Plate Vial 700 μL Taper
60-105-425 Square Well Plate Vial 1000 μL Round
60-105-403 Microtiter Plate Vial 1100 μL Round
60-105-405 Deep Well Plate Vial 1500 μL Round
03-252-446 Mat Applicator Not Applicable Not Applicable
03-252-445 Sealing Tape Applicator Not Applicable Not Applicable
60-105-432 Tube Tray Not Applicable Not Applicable
60-105-427 Center Hole Cap Not Applicable Not Applicable
60-105-429 Microtiter Plate 350 μL Flat Bottom (F-well)
60-105-406 Square Well Plate Vial 1000 μL Square
60-105-416 Microtiter Plate 2200 μL Round Bottom (U-well)
60-105-434 Automated Sample Concentrator Not Applicable Not Applicable
60-105-435 Needle Kit with Fitting Tool Not Applicable Not Applicable
60-105-431 Block Not Applicable Round Bottom (U-well)
60-105-415 Rack Not Applicable Not Applicable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 60-105-402 Supplier Thermo Scientific™ Supplier No. 60180V103

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Effectively seal all microplate formats to minimize evaporation and protect samples from contamination and spilling, ensure a consistent, reliable seal with all microplate formats with this easy-to-use, handheld applicator, and get significantly quicker dry-down times than standard methods, and remove most chromatography solvents in just minutes with easy to use 96-well sample concentrators.

Sealing Tape Applicator
Ensure a consistent, reliable seal with all microplate formats and all types of tape with this easy to use, handheld applicator for Thermo Scientific™ WebSeal Sealing Tapes. They are packaged two per box.

Sample Concentrators
Remove most chromatography solvents in just minutes. Simple to operate and maintain, Thermo Scientific™ WebSeal Sample Concentrators allow quick, easy solvent evaporation from 96 well microplates, offering significantly quicker dry-down times than standard methods such as vacuum ovens or centrifuges. These evaporators offer significant throughput advantages for laboratories looking to accelerate microplate sample preparation. Depending on your throughput requirements, choose either the manually operated MiniVap Concentrator or the programmable, robot-ready UltraVap.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Packaging 100 Pk.
Product Type Microtiter Plate Vial
For Use With (Equipment) 96-Well Microtiter Plate
Material Silicone, PTFE
No. of Wells 96
Volume (Metric) Working 700 μL
Total Volume (Metric) Well 700 μL
Well Shape Round
No. per Pack 100
Plate Compatibility 96-well Microtiter Plates
Quantity 100/Pk.
Height (Metric) 41.4 mm
Outside Diameter (Metric) 6.5 mm
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.