Learn More
Accumax Lab Devices Well Plate Sealing Membrane
Catalog No. 02945147
Change view
Click to view available options
Material:
Aluminum
Catalog No. Material
02-945-147 Aluminum
1 options
Catalog No. 02-945-147 Supplier Accumax Lab Devices Supplier No. 335813
Add to Cart
Add to Cart
These consumables are highly heat resistant up to 121°C thus making it compatible with every type of PCR reaction.
  • Easy adhesive film, no pressure required while applying
  • Strong adhesive, prevents evaporation during thermal cycles
  • Temperature stability range from -20 to 120°C
  • Provides supreme evaporation protection
  • Easy to pierce for sample recovery
  • Better protection for light sensitive samples
Specifications
Color Silver
Material Aluminum
Sterility Sterile
Autoclavable Autoclavable
No. per Case 100
No. per Pack 10
Product Type Well Plate Seal
Adhesive Adhesive Foil Seal
Pierceable Yes
Fit for Purpose PCR Ready