Learn More
Wells Lamont™ Split Back Grain Leather Drivers Gloves
Catalog No. 19148737 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-737 Medium
19-148-736 Small
19-148-738 Large
19-148-739 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-148-737 Supplier Wells Lamont™ Supplier No. Y0143M
Add to Cart
Add to Cart
This item is not returnable. View return policy
Split Back Grain Leather Drivers Gloves are constructed to combine quality and comfort to help reduce hand fatigue. Leather has a high level of tensile strength, and natural abrasion resistant properties that decrease hand lacerations.
  • Grain leather palm, split back
  • Double shirred elastic wrist
  • Bound hem
  • Keystone thumb
  • Gunn cut
Specifications
Cuff Material Elastic
Color Natural
Hand Specific
Material Grain Cowhide Leather
Product Type Driver Gloves
Size Medium
Style Gunn
Glove Cuff Style Open
Hem Color Green
Palm Type Grain Leather
Thumb Style Keystone