Learn More
Specifications
Specifications
| Color | Gray, Yellow |
| Cut Resistance Rating | ANSI A4 |
| Material | Stainless Steel Fiber, Polyester, Leather |
| Product Type | Cut-resistant Gloves |
| Size | Medium |
| For Use With (Application) | High performance metal working and general industrial applications |
| Glove Cuff Color | Green |
| Hem Color | Green |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.