Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Cut Reistant Gloves

Catalog No. 19148872 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
Hem Color:
Blue
Brown
Green
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hem Color
19-148-872 Large Brown
19-148-871 Medium Green
19-148-873 X-Large Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-148-872 Supplier Wells Lamont™ Supplier No. 1790L
This item is not returnable. View return policy

Specifications

Color Gray, Yellow
Cut Resistance Rating ANSI A4
Material Stainless Steel Fiber, Polyester, Leather
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) High performance metal working and general industrial applications
Glove Cuff Color Brown
Hem Color Brown
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.