Learn More
Wells Lamont™ Cut-Resistant FR Sleeves
GSA_VA
Catalog No. 19148973 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Style:
Sleeve with Thumb Hole
Standard Sleeve
Length (English):
10 in.
14 in.
18 in.
24 in.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style Length (English)
19-148-973 Standard Sleeve -
19-148-971 Standard Sleeve 14 in.
19-148-972 Standard Sleeve 18 in.
19-148-974 Sleeve with Thumb Hole 18 in.
19-148-975 Sleeve with Thumb Hole 24 in.
19-148-970 Standard Sleeve 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-148-973 Supplier Wells Lamont™ Supplier No. SKC24
Add to Cart
Add to Cart
This item is not returnable. View return policy
Composite Cut-Resistant FR Sleeves offer a high level of cut protection, while also providing outstanding comfort and fit. Made of a combination of high performance materials and flame resistant fibers, which will allow it to be used where incidental protection against flame and sparks are required.
  • Unique knit sleeve made from a patented combination of high performance materials and flame resistant fibers
  • Knitted using strong para-aramid yarns
  • Lightweight construction will increase employee acceptance and reduce the risk of injury
  • Blousy fit allows superior comfort and breathability
  • Elastic on both ends allows sleeve to stay in place
  • Available in several lengths, with or without a thumb hole, and also a wide top design for larger biceps
  • ANSI Cut Level A3
  • ANSI Flame Rating 4
  • Sold by the dozen pieces
  • Made in the USA
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Cut-resistant Sleeve
Style Standard Sleeve
Closure Type Elastic Ends
Length (Metric) 60.96 cm