Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Double Palm Shoulder Split Cowhide Leather Gloves

Catalog No. 19148801 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-801 Large
19-148-800 Small
19-148-802 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-148-801 Supplier Wells Lamont™ Supplier No. Y3101L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

An ideal glove for heavy duty abrasive environments that require additional protection in the palm and forefinger. The leather knuckle strap provides extra protection for the back of the hand, while the canvas back increases air circulation.

  • Select shoulder split cowhide leather construction
  • Double palm and forefinger
  • Striped back and cuff
  • Wing thumb
  • Safety cuff

Specifications

Back Material Canvas
Color Gray, Blue
Grade Select
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type Double Palm Gloves
Size Large
Glove Cuff Color Blue with Yellow Stripes
Glove Cuff Style Safety
Hem Color Brown
Series Y3101
Thumb Style Wing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.