Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Y9287 Polyurethane Coated Nylon Work Gloves

Catalog No. 19148863 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-863 Large
19-148-861 Small
19-148-862 Medium
19-148-864 X-Large
19-148-865 3X-Large
19-148-845 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-148-863 Supplier Wells Lamont™ Supplier No. Y9287L
Add to Cart
Add to Cart
This item is not returnable. View return policy

FlexTech™ Y9287 Polyurethane Coated Nylon Work Gloves are ideal for a variety of applications that require cut protection, along with high levels of dexterity. The glove's non-slip surface allows workers to work efficiently with confidence.

  • Synthetic seamless knit shell is comfortable and breathable
  • Polyurethane palm coating offers tactility and enhances grip
  • Provides additional protection to high wear/high risk areas of the hand
  • Dark coloring hides dirt and grime to extend the glove's useful life

Specifications

ANSI Abrasion Rating ANSI 4
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Electronic Assembly, and Small Parts Handling
Glove Cuff Style Knit
Series Y9287
Yarn Gauge 13
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.