Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Latex Coated Polyester/Cotton String Knit Gloves

Catalog No. 19148706 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-706 Medium
19-148-705 Small
19-148-704 X-Small
19-148-707 Large
19-148-708 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-148-706 Supplier Wells Lamont™ Supplier No. Y9240MVP

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Gloves feature a string knit cotton polyester shell, with a latex coating on the palm for enhanced grip and abrasion resistance.

  • Gray polyester/cotton string-knit shell with blue latex palm coating
  • Latex coating enhances grip capabilities, as well as abrasion/puncture resistance
  • Gray color conceals dirt
  • Flexible and comfortable
  • Packed in individual polybags

Specifications

Coated Area Palm
Construction String Knit
Coating Material Latex
Color Blue, Gray
Exterior Finish Partially Textured
Hand Specific
Material Polycotton
Size Medium
For Use With (Application) Canning, Recycling, and General Manufacturing
Glove Cuff Style Knit
Series Y9240-VP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.