Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Y9275 Polyurethane-Coated HPPE Cut-Resistant Gloves

Catalog No. 17200315 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-315 X-Large
17-200-311 X-Small
17-200-312 Small
17-200-313 Medium
17-200-314 Large
17-200-316 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-200-315 Supplier Wells Lamont™ Supplier No. Y9275XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

FlexTech™ Y9275 Polyurethane-Coated HPPE Cut-Resistant Gloves with High Performance Polyethylene (HPPE) fibers provide cut protection, while polyurethane coating on the palm/fingers enhances grip and tactility.

  • FlexTech high performance polyethylene fibers (HPPE) and nylon blended shell
  • Gloves provide a balance of comfort, dexterity and cut resistance
  • Polyurethane palm coating enhances grip and tactility
  • Ideal for general assembly and other applications where cut resistance is required
  • Launderable for a longer wear life

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance ASTM F1790-97, ASTM F2992-15, ASTM D3389-10
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Hand Specific
Material HPPE
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard EN 388:4342
For Use With (Application) Auto assembly, engine assembly, construction, metal fabrication
Glove Cuff Style Knit
Model Y9275XL
Puncture Rating ANSI/ISEA 105-16 Level 3
Series FlexTech™ Y9275
Yarn Gauge 13
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

WARNING: Cancer – www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.