Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Y9290 Full NBR-Coated Cut-Resistant Gloves with Nitrile-Coated Palm

Catalog No. 17200301 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-301 Medium
17-200-300 Small
17-200-302 Large
17-200-303 X-Large
17-200-304 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-200-301 Supplier Wells Lamont™ Supplier No. Y9290M
Add to Cart
Add to Cart
This item is not returnable. View return policy

FlexTech™ Y9290 Full NBR-Coated Cut-Resistant Gloves with Nitrile-Coated Palm is a lightweight, cut resistant shell that is double coated with a layer of nitrile rubber (NBR) for water-resistance, and has an additional layer of nitrile coating on the palm and fingers for cut protection.

  • Lightweight, cut-resistant knit shell with double coating
  • Sandy nitrile coated palm and fingertips enhances grip
  • Full NBR coating provides an additional layer of protection against liquids
  • Gloves offer a balance of comfort, dexterity and cut resistance
  • Water resistant

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ASTM F1790-97, ASTM D3389-10
Coated Area Palm, Fingertips
Construction Knit
Coating Material Nitrile
Color Gray
Cut Resistance Rating ANSI A4
Exterior Finish Fully Textured
Hand Specific
Material Nitrile Rubber
Product Type Cut-resistant Gloves
Size Medium
EN388 Standard EN 388:4543
For Use With (Application) Construction, metal fabrication, assembly
Glove Cuff Color Green
Glove Cuff Style Knit
Model Y9290M
Puncture Rating ANSI/ISEA 105-16 Level 4
Series Flextech™ Y9290
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.