Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Polyurethane Palm

Catalog No. 19148797 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Glove Cuff Color:
Black
Blue
Brown
Green
Red
Yellow
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Glove Cuff Color Size
19-148-797 Red Small
19-148-740 Yellow X-Small
19-148-796 Green Medium
19-148-798 Brown Large
19-148-831 Blue X-Large
19-148-832 Black 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-148-797 Supplier Wells Lamont™ Supplier No. Y9284S

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Fit and comfortable protection

  • Features highly cut-resistant fiber that protects against lacerations
  • Polyurethane palm coating offers grip and abrasion resistance
  • Dark-colored shell conceals dirt and grime
  • Knit wrist construction prevents dirt and debris from entering the glove
  • Color: Gray

Specifications

Color Gray
Cut Resistance Rating ANSI A3
Material Polyurethane
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Protection from lacerations and superior resistance against cuts and for grip and abrasion resistance
Glove Cuff Color Red
Glove Cuff Style Knit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.