Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Spec-Tec™ Cut Resistant Glove Liners
GSA_VA

Catalog No. 19044165 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-044165 Large
19-044162 Small
18-044-163 Medium
19-044166 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-044165 Supplier Wells Lamont™ Supplier No. M114L
Add to Cart
Add to Cart
This item is not returnable. View return policy

HPPE fibers are comfortable and stretchy, while also providing ANSI Cut Level A2 protection. Spec-Tec™ Cut Resistant Glove Liners can be laundered up to 12 times without shrinking.

Lightweight and comfortable

  • Constructed from a blend of Lycra™ and HPPE fibers
  • Liners provide ANSI Cut Level A2 protection
  • Launderable up to 12 times without shrinking
  • Latex-free
  • Made in the USA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A2
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material HPPE, Lycra
Product Type Cut-resistant Glove Liners
Size Large
Sterility Non-sterile
For Use With (Application) Organ/Tissue Recovery, Pharmaceuticals, and Hospitals
Glove Cuff Style Knit
Hem Color Blue
Series M114
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.