Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Whizard™ Silver Talon™ Cut-Resistant Gloves

Catalog No. 17200410 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-410 X-Small
19-050-742 Small
19-055-131 Medium
19-055-132 Large
19-055-133 X-Large
17-200-409 2X-Small
17-200-411 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-200-410 Supplier Wells Lamont™ Supplier No. 134525
Add to Cart
Add to Cart
This item is not returnable. View return policy

ANSI Level A7 Whizard™ Silver Talon™ Cut-Resistant Gloves are CFR 21 approved for food contact and are also low lint.

  • ANSI Level A6 cut resistant
  • High level of cut resistance
  • CFR 21 approved for food contact
  • Ambidextrous
  • 10-gauge thickness
  • Category III for particulates
  • Made in the USA

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance FDA CFR 21, ASTM F2992-15, ASTM D3884-09
Coating Material Uncoated
Color Silver, Blue
Cut Resistance Rating ANSI A5
Hand Ambidextrous
Latex Free Yes
Material Fibers, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size X-Small
EN388 Standard EN 388:X441
For Use With (Application) Meat processing industry
Glove Cuff Color Blue
Glove Cuff Style Knit
Model 134525
Series Whizard™ Silver Talon™
Yarn Gauge 10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.