Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Cut Resistant Glove Liners

Catalog No. 19048677 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048-677 X-Small
19-050-033A Small
19-050-033B Medium
19-050-033C Large
19-050-033D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-048-677 Supplier Wells Lamont™ Supplier No. M214XS
This item is not returnable. View return policy

Lightweight, thin glove liners are highly reusable and provide ANSI A1 cut resistance.

  • Lightweight, thin, and highly reusable white glove liner
  • Provides ANSI Cut Level A1 protection
  • Category II for particulates
  • Can be used alone or as a glove liner
  • Ambidextrous
  • Made in the USA

Specifications

Color White
Cut Resistance Rating ANSI A1
Disposable Reusable
Hand Ambidextrous
Material HPPE
Product Type Cut-resistant Glove Liners
Size X-Small
For Use With (Application) Cleanroom, Medical, or Semi-Conductor Industries
Glove Cuff Style Knit
Hem Color Yellow
Series M214
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.