Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Standard Shoulder Split Leather Palm Gloves - Gauntlet Cuff

Catalog No. 1146238 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-462-38 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-462-38 Supplier Wells Lamont™ Supplier No. Y3202L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

This durable glove offers split leather index fingers, leather tips, and a leather knuckle strap. The lined palm provides added comfort in the palm of the hand.

  • Standard shoulder split cowhide leather palm
  • Full leather palm, fingertips and knuckle strap
  • Striped canvas back and cuff
  • Wing thumb
  • Gauntlet cuff
  • Lined palm

Specifications

Back Material Canvas
Color Gray, Red
Grade Standard
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type Leather Palm Gloves
Size Large
Style Gunn
Glove Cuff Color Red Stripes
Glove Cuff Style Gauntlet
Hem Color Brown
Thumb Style Wing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.