Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Thermofill™ Lined Leather Palm Gloves

Catalog No. 19148750 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-750 Large
19-148-749 Medium
19-148-751 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-148-750 Supplier Wells Lamont™ Supplier No. Y0042L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pigskin Leather Palm Gloves provide comfort and thermal protection with Thermofill™ lining. The knuckle strap and safety cuff provide additional protection for the knuckles and wrist.

  • Premium pigskin leather palm
  • Thermofill™ lined
  • Safety cuff
  • Wing thumb
  • Gunn cut

Specifications

Cuff Material Fabric
Color Tan
Grade Premium
Hand Specific
Material Pigskin Leather
Product Type Thermal Lined Gloves
Size Large
Style Gunn
For Use With (Application) Ideal in the outdoors, freezers or any cold environments.
Glove Cuff Color Blue with Yellow Stripes
Glove Cuff Style Safety
Hem Color Brown
Liner Material Thermofill™
Series Y0042
Thumb Style Wing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.