Learn More
Wells Lamont™ MT130 Sterile/Non-Sterile Touchscreen Cut Resistant Gloves
Shop All Wells Lamont Industry Group ProductsDescription
- Patent pending yarn technology delivers cut resistance and touchscreen capability
- Minimal particle shedding to meet cleanroom classifications and requirements
- Meets Category II using Helmke Drum Test referenced in IEST –RP-CC003.3
- ANSI Level 4
- Ambidextrous gloves available in different sizes
Specifications
Specifications
| Color | Gray |
| Product Type | Cut-resistant Gloves |
| Size | X-Small |
| Glove Cuff Color | Gold |