Learn More
ProteinSimple Anti-Rabbit Detection Module, 1ea
Supplier:  ProteinSimple DM001
Encompass
Use with a Wes or Peggy Sue/Sally Sue Separation Module. Includes: Luminol-S, Peroxide, Antibody Diluent 2, Anti-Rabbit Secondary Antibody, Streptavidin-HRP.
Catalog No. NC1196891