Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester™ Acrylic/Polyester General Purpose Gloves with PVC Grip

Catalog No. 19821555 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-821-555 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-821-555 Supplier West Chester™ Supplier No. 708SKH/M
This item is not returnable. View return policy

Criss-cross pattern PVC for increased grip and abrasion resistance.

  • Orange acrylic and polyester string knit
  • Orange PVC crisscross grip pattern

Specifications

Construction String Knit
Cuff Material Elastic
Coating Material PVC
Color Orange
Hand Ambidextrous
Material Polyester, Acrylic
Product Type General Purpose Gloves
Size Medium
Glove Cuff Color Orange
Glove Cuff Style Knit
Hem Color Green
Lining Seamless Lining
Palm Type PVC Grip
Length (English) 9.25 in.
Length (Metric) 23.5 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

WARNING: California Proposition 65 - Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.