Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Economy Polypropylene Lab Coat

Catalog No. 191301293E Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-1293E 2X-Large
19-130-1293F 3X-Large
19-130-1293D X-Large
19-130-1293C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-1293E Supplier West Chester Supplier No. 3514/XXL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Standard weight polypropylene lab coat protects against dry particulates and worn when full coverage is not necessary.

  • Standard weight spunbond polypropylene
  • Chest and hip pockets
  • Snap buttons for quick donning
  • Professional collar
  • Single-use, white

Specifications

Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
Resistance Dust
Style Standard Length
No. of Pockets 2
Latex Free Yes
Model 3514
Seam Type Serged
For Use With (Application) Manufacturing, Pharmaceuticals, Cleaning, Laboratories
Construction Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.