Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Flock lined Unsupported Gloves

Catalog No. 19821537 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Flock lined gloves; Length: 13 in.; 28 mil; Black; X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-821-537 Flock lined gloves; Length: 13 in.; 28 mil; Black; X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-821-537 Supplier West Chester Supplier No. 32212/XL
This item is not returnable. View return policy

Specifications

Color Black
Latex Free Yes
Material Neoprene
Size X-Large
Thickness (English) 28 mil
For Use With (Application) Food service, janitorial, hazardous cleanup, chemical handling
Glove Cuff Style Beaded/Rolled
Lining Lined
Length (English) 13 in.
Length (Metric) 33 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.