Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Heavy Weight Spunbound Polypropylene Coveralls

Catalog No. 19821611 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Large
4X-Large
5X-Large
Large
Medium
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-821-611 Medium
19-821-610 Large
19-821-609 2X-Large
19-821-608 4X-Large
19-821-612 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-821-611 Supplier West Chester Supplier No. U1200/M
This item is not returnable. View return policy

Providing barrier protection against non-hazardous dry particulates

  • Made from polypropylene material
  • Full length zipper front for easy donning
  • Elastic Wrist and Ankles keeps dirt and debris out
  • High level of comfort and breathability. Cost effective protection
  • Color: White
  • Available in sizes M - 5XL
  • Used for worker safety in a wide variety of industrial applications including general labor, laboratory, pharmaceutical, cleaning, and janitorial applications

Specifications

Product Type General Purpose Coveralls
Size Medium
Material Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Closure Type Front Zipper (Full-length)
Sterility Non-sterile
Latex Free Yes
For Use With (Application) General Labor, Laboratory, Pharmaceutical, Cleaning and Janitorial
Style Regular
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.