Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Chemical Resistant 3312 Latex Gloves, Flock Lining

Catalog No. 19050143D Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size: 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-050-143D Size: 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-050-143D Supplier West Chester Supplier No. 3312/10
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Economy yellow latex gloves are chemical, puncture and tear resistant.

  • Flock lined latex
  • 16 mil
  • 12 in. length
  • USDA accepted for use in meat and poultry applications
  • Applications include maintenance, general labor, janitorial, food service

Specifications

Color Yellow
Latex Free No
Material Latex
Size 10
Thickness (English) 16 mil
Model 3312/10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.