Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester™ Striped Canvas Gloves with Leather Patch Palm/Fingers

Catalog No. 19821531 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-821-531 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-821-531 Supplier West Chester™ Supplier No. 100P
This item is not returnable. View return policy

Uncoated gloves with a 5-piece canvas back provide great protection and finger dexterity. Knit wrist helps keep dirt and debris out of the glove.

  • Leather patch palm and fingers
  • 5-piece, red/blue striped canvas back

Specifications

Color Red, Blue
Grade Industrial
Hand Specific
Material Cotton, Canvas
Product Type General Purpose Gloves
Size Large
Gender Men's
Glove Cuff Color Natural
Glove Cuff Style Knit
Liner Material Cotton/Polyester Blend
Palm Type Cowhide Leather
Length (English) 10.3 in.
Length (Metric) 26 cm
Show More Show Less

WARNING: California Proposition 65 - Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.