Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester™ Top Grain Pigskin Leather Drivers Gloves with Red Fleece Lining

Catalog No. 19821572 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-821-572 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-821-572 Supplier West Chester™ Supplier No. 9940KF/L
This item is not returnable. View return policy

Top grain pigskin leather construction provides breathability due its porous texture, becomes softer with use, and withstands moisture without stiffening. Ideal for use in cold weather applications including farming, construction, machine operation, and refrigeration.

  • Top grain leather is smooth, provides durability and dexterity
  • Red fleece lining provides insulation
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Keystone thumb is ergonomic, reduces hand fatigue, offers dexterity, and durability
  • Elastic closure sewn inside glove for safety and comfort
  • Color coded hems for easy size identification

Specifications

Color Gold
Grade Premium
Hand Specific
Material Top Grain Pigskin Leather
Product Type Driver Gloves
Size Large
Style Gunn
For Use With (Application) Construction, Masonry, Agriculture, Iron/Steel Work, Mining, Foundry Operations, Transportation, and Electrical Work
Glove Cuff Style Open
Liner Material Fleece
Series 9940KF
Thumb Style Keystone
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.