Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester PE Laminate Basic Coveralls

Catalog No. 190882268A Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Light-weight; Serged seam; White; Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-088-2268A Light-weight; Serged seam; White; Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-088-2268A Supplier West Chester Supplier No. 3400/L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Size Large
Material Polyethylene, Polypropylene
Color White
Ankle Style Open
Cuff Style Open
Closure Type Zipper
Seam Type Serged
Description Full length zipper front, open wrist and ankle
For Use With (Application) Manufacturing, mold remediation, cleaning, abatement
Length (English) 37 in.
Length (Metric) 93.98 cm
Width (English) Chest 25 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.