Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Polypropylene Disposable Beard Covers

Catalog No. 19068442 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Quantity:
100/Cs.
1000/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-068-442 1000/Cs.
19-121-070 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-068-442 Supplier West Chester Supplier No. UBC1000
  • $281.00
    $221.65 / Case of 1000

    Save $59.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Polypropylene disposable beard cover provides protection against non-hazardous dry particulates.

  • One size fits most
  • Elastic band for flexible fit
  • Latex free
  • White
  • 1000 count
  • Made from component materials that comply with Federal Regulations for Food Contact: 21CFR 170-199

Specifications

Size One Size
Material Spunbond Polypropylene
Color White
Disposable Single-use
Latex Free Yes
For Use With (Application) Food Industry, Light Maintenance
Quantity 1000/Cs.
Length (English) 18 in.
Length (Metric) 45.72 cm
Product Type General Purpose Beard Cover
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.