Learn More
West Chester Posi M3 Sleeves
Catalog No. 19055158 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Universal
Catalog No. Size
19-055-158 Universal
1 options
This item is not returnable. View return policy
Catalog No. 19-055-158 Supplier West Chester Supplier No. C3812
  • $286.00
    $218.65 / Case of 200
    Save $67.35
    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Five layers of protection providing a high level of breathability and protection against dry particulates and water based light liquid sprays and splashes. Good for use in industrial manufacturing, paper industry, food processing and any heavy dust environment.
Five layers of protection For use in industrial manufacturing, paper industry, food processing and any heavy dust environment
Specifications
Size Universal
Product Type General Purpose Sleeve
Material Polyethylene, Spunbond Polypropylene
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Serged
For Use With (Application) Industrial manufacturing, paper industry, food processing
Length (Metric) 45.7 cm