Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Posi-Wear™ BA Coveralls

Catalog No. 19821553 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-821-553 Large
19-821-554 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-821-553 Supplier West Chester Supplier No. 3609/L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Coveralls provide five layers of protection against dry particulates and water based light liquid sprays and splashes.

For use in manufacturing, pressure washing, painting, resurfacing or fiberglass handling.

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Protection Class None
Resistance Dust
Latex Free Yes
No. of Pockets None
For Use With (Application) Agriculture, general industry, construction and municipalities
Style Regular
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.