Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester™ PVC Coated Cotton Work Gloves with Textured Finish

Catalog No. 19821577 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-821-577 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-821-577 Supplier West Chester™ Supplier No. J1027RF
  • $185.00
    $93.65 / Dozen of 12

    Save $91.35

    49% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $965.00
    $488.65 / Case of 6 DZ

    Save $476.35

    49% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are fully coated with PVC to provide abrasion resistance in addition to protection from grease, oil, and acid.

  • Fully coated PVC gloves provide protection from acid, oil, and grease
  • Cotton jersey liner increases user comfort

Specifications

Coating Material PVC
Color Black
Exterior Finish Partially Textured
Hand Specific
Latex Free Yes
Material PVC
Product Type Dip-coated Gloves
Size Large
For Use With (Application) General Maintenance, and Utility Work
Glove Cuff Style Gauntlet
Liner Material Cotton
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.