Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester™ Select Brushed Pigskin Palm Gloves

Catalog No. 19821548 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-821-548 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-821-548 Supplier West Chester™ Supplier No. 500P
This item is not returnable. View return policy

Pigskin leather construction provides breathability due its porous texture. Gloves become softer with use and withstand moisture without stiffening.

  • Gunn cut pattern delivers durability, wear, and comfort
  • Wing thumb increases comfort in open-handed work
  • Fabric back provides breathability and flexibility
  • Leather knuckle strap and finger tips for extra protection
  • Elastic closure sewn inside glove for safety and comfort
  • Safety cuff helps protect wrist and allows for fast removal
  • Ideal for pipe, brick & other material handling, construction, farming and general maintenance

Specifications

Cuff Material Rubberized
Color Tan, Blue
Grade Select
Hand Specific
Material Pigskin Leather
Product Type Leather Palm Gloves
Size Large
Style Gunn
Glove Cuff Color Blue with Red Stripes
Glove Cuff Style Safety
Thumb Style Wing
Length (English) 10 in.
Length (Metric) 25.4 cm
Weight 50.7 lb. (Case)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.