Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester™ Split Cowhide Leather Double Palm Gloves

Catalog No. 189992766 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-999-2766 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-999-2766 Supplier West Chester™ Supplier No. 800DP
This item is not returnable. View return policy

Gloves feature reinforced double split leather on the palm and index finger, a rubberized safety cuff, and a pull tab at the cuff for easy removal.

  • Double palms offers comfort, insulation and heat protection
  • Gunn cut pattern delivers durability, wear, and comfort
  • Wing thumb increases comfort in open-handed work
  • Fabric back provides breathability and flexibility
  • Leather knuckle strap and finger tips for extra protection
  • Elastic closure sewn inside glove for safety and comfort
  • Rubberized gauntlet cuff for added forearm protection, allows for fast removal and water resistance
  • Used for construction, material handling, and general maintenance

Specifications

Back Material Canvas
Cuff Material Rubberized
Color Gray, Green
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type General Purpose Gloves
Size Large
Style Gunn
Glove Cuff Color Green with Pink Stripes
Glove Cuff Style Gauntlet
Hem Color Brown
Thumb Style Wing
Length (English) 12 in.
Length (Metric) 30.5 cm
Weight 51.7 lb. (Case)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.